Cyclohexylacetic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Cyclohexaneacetic acid

C0589-25G C0589-250G
TCIAC0589-25GEA 795 NOK
TCIAC0589-25G TCIAC0589-250G
Cyclohexylacetic acid ≥98.0% (by GC, titration analysis)
Cyclohexylacetic acid
Formel: C₈H₁₄O₂
MW: 142,2 g/mol
Kokepunkt: 242 °C (1013 hPa)
Smeltepunkt: 26…27 °C
Tetthet: 0,9985 g/cm³ (22 °C)
MDL nummer: MFCD00001518
CAS-nummer: 5292-21-7
EINECS: 226-132-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5ae1e24b9d048648709ab37dfdb3c4ef3f4631b&vid=d5ae1e24b9d048648709ab37dfdb3c4ef3f4631b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR