(S)-(-)-Tetrahydro-2-furoic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: (S)-Tetrahydrofuran-2-carboxylic acid, (2S)-(-)-Tetrahydrofuran-2-carboxylic acid

T1741-1G T1741-25G T1741-5G
TCIAT1741-1GEA 220 NOK
TCIAT1741-1G TCIAT1741-25G TCIAT1741-5G
(S)-(-)-Tetrahydro-2-furoic acid ≥98.0% (by GC, titration analysis)
(S)-(-)-Tetrahydro-2-furoic acid
Formel: C₅H₈O₃
MW: 116,12 g/mol
Flash punkt: 139°C
MDL nummer: MFCD00272639
CAS-nummer: 87392-07-2
FN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eea230ddbe1b563ac429e115cafaac86db6b4b3&vid=3eea230ddbe1b563ac429e115cafaac86db6b4b3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -18.0 to -20.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR