Digital pocket refractometers for food, PAL series
Leverandør: Atago
4427 4429 4430 4439 4498
635-0234EA 4150 NOK
635-0234 635-0235 635-0236 635-0243 ATAG4498
Digital pocket refractometers for food, PAL series
Refraktometere Refraktometere, håndholdte
Battery operated instruments for food applications.
  • Single handed operation
  • Easy to use and delivering rapid results
  • Corresponding temperature range also displayed
Sertifiseringer: IP 65 for rinsing off the sample with running water