Handheld refractometers for wine
Leverandør: Atago
4484 4479 4480 4483 4486 4487
635-0157EA 4150 NOK
635-0157 635-0277 635-0278 635-0279 635-0281 635-0282
Handheld refractometers for wine
Refraktometere Refraktometere, håndholdte
The PAL series with special scales are pocket sized.
  • Simple handed operation
  • Rapid results: Three second response
  • Battery operated instruments
Sertifiseringer: IP 65 allows rinsing off the sample with running water.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Prøvestørrelse 0,3 ml
Responstid 3 seconds
Temperaturkompenseringområde 0…40 °C
Elektronisk Two AAA batteries
B×D×H 55×31×109 mm
Vekt 100 g