Dual/Triple scale handheld refractometers, MASTER series
Leverandør: Atago
2771 2773 2791 2793
ATAG2771EA 3900 NOK
ATAG2771 ATAG2773 ATAG2791 ATAG2793
Dual/Triple scale handheld refractometers, MASTER series
Refraktometere Refraktometere, håndholdte
The MASTER refractometers are designed for clinical applications.
  • Plastic design resistant to salty and acid samples
  • The (alpha) models include ATC and are IP 65 rated except eyepiece
  • Ergonomic grip and prism housing
Samples cover the prism without having to lift the daylight plate.
Bestill nå
SPESIFIKASJONER
Modell MASTER-URC/Nα MASTER-URC/NM MASTER-URC/Nα MASTER-URC/NM
Environmentally Preferable
Prøvestørrelse 1 drop 1 drop 1 drop 1 drop
Nøyaktighet ±0,0005 nD ±0,0005 nD ±0,001 nD ±0,001 nD
område 1,3330 - 1,3660 (nD) 1,3330 - 1,3660 (nD) 1,333 - 1,356 (nD) 1,333 - 1,356 (nD)
Oppløsning 0,001 SG/0,0005 nD/ 0,2 g/100 ml 0,001 SG/0,0005 nD/ 0,2 g/100 ml 0,001 SG/nD 0,001 SG/nD
Vekt 110 g 110 g 110 g 110 g