UHPLC columns, Pinnacle® DB Aqueous C18 (USP L1)
Supplier: Restek
9418212 9418232 9418252 941850210 941850212 9418312 9418312-700 9418313-700 9418315 9418315-700 941831E 9418332 9418332-700 9418333-700 9418335 9418335-700 941833E 9418352 9418352-700 9418353-700 9418355 9418355-700 941835E 9418362 9418362-700 9418363-700 9418365 9418365-700 941836E 9418512 9418512-700 9418513-700 9418515 9418515-700 941851E 9418522 9418522-700 9418523-700 9418525 9418525-700 941852E 9418532 9418532-700 9418533-700 9418535 9418535-700 941853E 9418552 9418552-700 9418553-700 9418555 9418555-700 941855E 9418562 9418562-700 9418563-700 9418565 9418565-700 941856E 9418572 9418572-700 9418573-700 9418575 9418575-700 941857E
554-1564EA 16075 NOK
554-1564 554-1565 554-1566 REST941850210 REST941850212 REST9418312 REST9418312-700 REST9418313-700 REST9418315 REST9418315-700 REST941831E REST9418332 REST9418332-700 REST9418333-700 REST9418335 REST9418335-700 REST941833E REST9418352 REST9418352-700 REST9418353-700 REST9418355 REST9418355-700 REST941835E REST9418362 REST9418362-700 REST9418363-700 REST9418365 REST9418365-700 REST941836E REST9418512 REST9418512-700 REST9418513-700 REST9418515 REST9418515-700 REST941851E REST9418522 REST9418522-700 REST9418523-700 REST9418525 REST9418525-700 REST941852E REST9418532 REST9418532-700 REST9418533-700 REST9418535 REST9418535-700 REST941853E REST9418552 REST9418552-700 REST9418553-700 REST9418555 REST9418555-700 REST941855E REST9418562 REST9418562-700 REST9418563-700 REST9418565 REST9418565-700 REST941856E REST9418572 REST9418572-700 REST9418573-700 REST9418575 REST9418575-700 REST941857E
UHPLC columns, Pinnacle® DB Aqueous C18 (USP L1)
Chromatography Columns
Highly selective phase for polar analytes. Compatible with highly aqueous (up to 100%) mobile phases. Silica manufactured by Restek.
Order Now