Protein quantitation assay kits, CB-X™ Protein assay
Leverandør: G Bioscienes
786-12X 786-12XT
786-12XEA 8950 NOK
786-12X 786-12XT
Protein quantitation assay kits, CB-X™ Protein assay
Assay Proteinanalyser
CB-X™ Protein Assay is a Coomassie dye based protein assay based on the Bradford Protein Assay. For protein samples in simple, uncomplicated aqueous buffers CB-X™ is a highly sensitive, single reagent assay that can be performed in 5 minutes. For complicated protein samples, CB-X™ Protein Assay is supplied with reagents to clean up the samples and remove all reagents and chemicals that interfere with accurate protein estimation in a single step. These reagents include detergents, chaotropes, reducing agents, alkylating agents, sugars, high salt concentrations, buffering agents and chelating agents. The clean up stage and subsequent protein assay is performed in a single tube to ensure no protein loss and to maintain the accuracy of the assay.
CB-X™ Protein Assay is supplied with lot specific CB-X™ Tables. These allow researchers to perform single protein clean ups, subsequent assays and then look up their absorbance in the CB-X™ Table to find the protein concentration. The CB-X™ Table eliminates the need for multiple protein standards and saves considerable time and effort. The CB-X™ Table is prepared with a complex protein mixture that compares well with proteins from mammalian, plant, bacteria and yeast sources. A set of bovine serum albumin standards are supplied for generating curves when using CB-X™ Assay Dye alone or for researchers who prefer to generate their own standard curve or to generate their own CB-X™ Table for their specific conditions.
The CB-X™ Protein Assay is reliable over the range of 0,5 to 50 μg per assay. Suitable for 500 protein assays and enough clean up reagents for 250 clean ups.