Water baths, unstirred, JB Academy series
Leverandør: GRANT INSTRUMENTS
JBA12 JBA18 JBA5
462-1019EA 10400 NOK
462-1019 462-1020 462-1021
Water baths, unstirred, JB Academy series
Bad Vannbad, uten sirkulasjon
Digital unstirred water bath range with choice of models from 5 to 18 litres. All units have stainless steel tanks and durable painted steel cases, offer fast heat-up times, good temperature control, front panel lock, easy to use controls and a bright, clear display. All baths have heater mat technology ensuring optimum working space, without a heating element in the tank they are easier to clean. The range is ideal for use in education and teaching as well as industrial applications such as product and materials testing.
  • Temperature stability ±0,5 °C, temperature range ambient 5 to 95