(-)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: (3aS,7aS)-Hexahydroisobenzofuran-1,3-dione

C1418-100MG C1418-1G
TCIAC1418-100MGEA 745 NOK
TCIAC1418-100MG TCIAC1418-1G
(-)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0% (by GC)
(-)-trans-1,2-Cyclohexanedicarboxylic anhydride
Formel: C₈H₁₀O₃
MW: 154,1632 g/mol
MDL nummer: MFCD00674195
CAS-nummer: 31982-85-1
EINECS: 238-009-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d38c964b854249122c308b0b62603754cb701abf&vid=d38c964b854249122c308b0b62603754cb701abf&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 164.0 to 168.0 deg-C
Specific rotation [a]20/D -80.0 to -90.0 deg (C=1, Dioxane)
Solubility in Dioxane almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR