2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester ≥97%
Leverandør: Thermo Scientific
Synonymer: 2-(2-(1,3-Dioxolan-2-yl)ethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H26071.03 H26071.06
H26071.03EA 985 NOK
H26071.03 H26071.06
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester ≥97%
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester
Purity: 97% Molecular Formula: C11H21BO4 Formula Weight: 228.10 Boiling Point: 100°/1.5mm MDL No.: MFCD03788722
Formel: C₁₁H₂₁BO₄
MW: 228,10 g/mol
Kokepunkt: 100 °C (1,5 mmHg)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03788722
CAS-nummer: 1073354-07-0