2,5-Dimethylbenzoxazole ≥98.0% (by GC)

Leverandør: TCI
Warning

D3461-25G D3461-5G
TCIAD3461-25GEA 2000 NOK
TCIAD3461-25G TCIAD3461-5G
2,5-Dimethylbenzoxazole ≥98.0% (by GC)
2,5-Dimethylbenzoxazole
Formel: C₉H₉NO
MW: 147,18 g/mol
Kokepunkt: 215°C
Flash punkt: 91°C
MDL nummer: MFCD00005773
CAS-nummer: 5676-58-4
EINECS: 227-136-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5166e0dc96388a63c52e02b5cefdee71e91c95f&vid=c5166e0dc96388a63c52e02b5cefdee71e91c95f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR