1H-Indazole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Indazole-3-carboxylic acid

I0672-25G I0672-5G
TCIAI0672-25GEA 1025 NOK
TCIAI0672-25G TCIAI0672-5G
1H-Indazole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1H-Indazole-3-carboxylic acid
Formel: C₈H₆N₂O₂
MW: 162,15 g/mol
Smeltepunkt: 261°C
MDL nummer: MFCD00211066
CAS-nummer: 4498-67-3
EINECS: 224-794-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fdcc12b4b80b06712fd2c08428a5d3128f9f10f&vid=4fdcc12b4b80b06712fd2c08428a5d3128f9f10f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR