(-)-Menthoxyacetic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)acetic acid, (-)-Menthyloxyacetic acid
M0573-25G
M0573-5G
TCIAM0573-25GEA
6350
NOK
TCIAM0573-25G
TCIAM0573-5G
(-)-Menthoxyacetic acid ≥98.0% (by titrimetric analysis)
(-)-Menthoxyacetic acid
Formel:
C₁₂H₂₂O₃ MW: 214,3 g/mol |
MDL nummer:
MFCD00001483 CAS-nummer: 40248-63-3 EINECS: 254-857-2 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Specific rotation [a]20/D | -92.0 to -96.0 deg (C=2, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...