5-Amino-2-methylbenzenesulphonamide 96%
Leverandør: Thermo Scientific
|
Warning
|
H34056.03
H34056.06
H34056.03EA
565
NOK
H34056.03
H34056.06
5-Amino-2-methylbenzenesulphonamide 96%
5-Amino-2-methylbenzenesulphonamide
Purity: 96%
CAS No.: 6973-09-7
Molecular Formula: C7H10N2O2S
Formula Weight: 186.23
Melting Point: 163-164°
MDL No.: MFCD06681071
Formel:
C₇H₁₀N₂O₂S MW: 186,23 g/mol Smeltepunkt: 163…164 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD06681071 CAS-nummer: 6973-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...