4-(Chloromethyl)benzoic acid 96%
Leverandør: Thermo Scientific
Danger
B22275.06 B22275.14
B22275.06EA 600 NOK
B22275.06 B22275.14
4-(Chloromethyl)benzoic acid 96%
4-(Chloromethyl)benzoic acid
Purity: 96% CAS No.: 1642-81-5 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Melting Point: 199-202° Beilstein Reference: 1907970 UN No.: 3261 MDL No.: MFCD00002568
Formel: C₈H₇ClO₂
MW: 170,60 g/mol
Smeltepunkt: 199…202 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002568
CAS-nummer: 1642-81-5
EINECS: 216-697-1
ADR: 8,III