Hydrogenklorid 1.25 mol/l in methanol, Supelco®
Leverandør: Merck
M-Clarity™ quality level = MQ100
17935-250ML 17935-50ML
17935-250MLEA 1160 NOK
17935-250ML 17935-50ML
Hydrogenklorid 1.25 mol/l in methanol, Supelco®
Hydrogenklorid
Formel: HCl
MW: 36,46 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09b8a4d46fc0d3f9d1cc4da5780d4eb4dee20bbc&vid=65e464419f65c4c7282c66bd2c43df2fdf049290&prid=web&clid=SAPDAM