4-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC)

Leverandør: TCI
Danger

B3120-1G B3120-5G
TCIAB3120-1GEA 785 NOK
TCIAB3120-1G TCIAB3120-5G
4-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC)
4-Bromo-2-hydroxybenzonitrile
Formel: C₇H₄BrNO
MW: 198,01 g/mol
MDL nummer: MFCD07780675
CAS-nummer: 288067-35-6
FN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca1531d92fe1e03fc1450d52770368eb1a66a639&vid=96ca1e016071df32fb118b94c8cf3b1263f3f1dd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR