Methyl-3,5-dibromo-4-methylbenzoate ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Methyl 3,5-dibromo-4-methylbenzoate
D2871-25G
D2871-5G
TCIAD2871-25GEA
1525
NOK
TCIAD2871-25G
TCIAD2871-5G
Methyl-3,5-dibromo-4-methylbenzoate ≥98.0% (by GC)
Methyl-3,5-dibromo-4-methylbenzoate
Formel:
C₉H₈Br₂O₂ MW: 307,98 g/mol |
MDL nummer:
MFCD00010729 CAS-nummer: 74896-66-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...