Methyl-4-bromo-3-methylbenzoate ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Methyl 4-bromo-3-methylbenzoate
B2256-25G
B2256-5G
TCIAB2256-25GEA
1575
NOK
TCIAB2256-25G
TCIAB2256-5G
Methyl-4-bromo-3-methylbenzoate ≥97.0% (by GC)
Methyl-4-bromo-3-methylbenzoate
Formel:
C₉H₉BrO₂ MW: 229,07 g/mol |
MDL nummer:
MFCD00673014 CAS-nummer: 148547-19-7 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...