1-Amino-2-methylbutane ≥85.0% (by GC) (contains 3-Methylbutylamine)

Leverandør: TCI
Danger

Synonymer: 2-Methylbutylamine, 2-Methyl-1-butylamine

M0767-25ML M0767-5ML
TCIAM0767-25MLEA 1275 NOK
TCIAM0767-25ML TCIAM0767-5ML
1-Amino-2-methylbutane ≥85.0% (by GC) (contains 3-Methylbutylamine)
1-Amino-2-methylbutane
Formel: C₅H₁₃N
MW: 87,16 g/mol
Kokepunkt: 97 °C (1013 hPa)
Tetthet: 0,755 g/cm³ (18 °C)
Flash punkt: 8°C
MDL nummer: MFCD00008147
CAS-nummer: 96-15-1
EINECS: 202-483-5
FN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62122da55c38d0d0d914da8ce132c9ea3ff6d19e&vid=62122da55c38d0d0d914da8ce132c9ea3ff6d19e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 85.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR