pH and ORP microelectrodes, ScienceLine / IoLine
Leverandør: SI Analytics
285105065 285105127 285105135 285105151 285105632 285105702 285105176
662-1672EA 8125 NOK
662-1672 662-1267 662-0520 662-6109 662-1676 662-1677 662-1673
pH and ORP microelectrodes, ScienceLine / IoLine
Elektroder pH-elektroder
For small sample volumes in life science and pharmaceutical applications.
  • Reliable and reproducible results
  • Highest quality and long lasting
  • Accurate and highly precise
  • Designed for small volumes and life science applications
Suitable temperature range: −5 to +100 °C.
pH electrodes are equipped with A-type membrane glass. pH electrodes with integrated temperature measurement are using a PT1000 sensor.
For most accurate results, we recommend using the ProLab 3000 meter in combination with these electrodes.