Ion exchange HPLC columns, BioBasic™ AX and SCX
Leverandør: Thermo Fisher Scientific
73205-154630 73105-254630 73205-254630 73205-104630 73205-053030 73205-014001 73105-154630 73105-152130 73105-054630 73105-053030 73105-052130 73105-012101 73205-052130 73205-101030 73205-054630 73205-251030 73205-152130 73205-151030 73205-102130 73105-153030 73105-151030 73105-051030 73105-104630 73105-102130 73105-101030
HYPE73205-154630EA 12475 NOK
HYPE73205-154630 HYPE73105-254630 HYPE73205-254630 HYPE73205-104630 HYPE73205-053030 HYPE73205-014001 HYPE73105-154630 HYPE73105-152130 HYPE73105-054630 HYPE73105-053030 HYPE73105-052130 HYPE73105-012101 HYPE73205-052130 HYPE73205-101030 HYPE73205-054630 HYPE73205-251030 HYPE73205-152130 HYPE73205-151030 HYPE73205-102130 HYPE73105-153030 HYPE73105-151030 HYPE73105-051030 HYPE73105-104630 HYPE73105-102130 HYPE73105-101030
Ion exchange HPLC columns, BioBasic™ AX and SCX
Kromatografikolonner
BioBasic™ AX LC columns optimized for the separation of proteins, peptides, other anionic species and highly polar molecules.
  • Low hydrophobicity
  • Use for normal phase and hydrophilic interaction chromatography (HILIC)
  • Weak anion exchange phase for multiple charged species
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 3% 3%
pH område 2 - 8 2 - 8
USP kode L52