Conjugation reagents, linkers
Leverandør: TRILINK BIOTECHNOLOGIES
S-1002-010 S-1002-105 S-1008-010 S-1037-010 S-1009-010 S-1001-010 S-1004-105 S-1008-105 S-1011-010 S-1004-010
733-2514EA 9600 NOK
733-2514 733-2513 733-2519 733-2520 733-1797 733-1884 733-1886 733-1888 SOLUS-1011-010 SOLUS-1004-010
Conjugation reagents, linkers
Assay Proteinanalyser