2-Methoxy-6-(tributylstannyl)pyrazine 95%
Leverandør: Thermo Scientific
|
Danger
|
H51528.MD
H51528.03
H51528.MDEA
755
NOK
H51528.MD
H51528.03
2-Methoxy-6-(tributylstannyl)pyrazine 95%
2-Methoxy-6-(tributylstannyl)pyrazine
Purity: 95%
CAS No.: 1105511-66-7
Molecular Formula: C17H32N2OSn
Formula Weight: 399.15
Storage Temperature: 2°C to 8°C
Formel:
C₁₇H₃₂N₂OSn MW: 399,15 g/mol |
MDL nummer:
MFCD08275735 CAS-nummer: 1105511-66-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...