10-Undecynoic acid 96%
Leverandør: Thermo Scientific
Warning
Synonymer: Undec-10-ynoic acid
A13815.06 A13815.14
A13815.06EA 1250 NOK
A13815.06 A13815.14
10-Undecynoic acid 96%
10-Undecynoic acid
Purity: 96% CAS No.: 2777-65-3 Molecular Formula: HC≡C(CH2)8CO2H Formula Weight: 182.26 Flash Point: 161°(321°F) Odour: Odourless Boiling Point: 174-176°/4mm Beilstein Reference: 1704918 MDL No.: MFCD00014389
Formel: C₁₁H₁₈O₂
MW: 182,26 g/mol
Kokepunkt: 174…176 °C (4 mmHg)
Smeltepunkt: 40…43 °C
Flash punkt: 161 °C (321 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014389
CAS-nummer: 2777-65-3
EINECS: 220-471-8