1H,1H,5H-Octafluoropentyl acrylate ≥97.0% (by GC) stabilised

Leverandør: TCI
Warning

Synonymer: 2,2,3,3,4,4,5,5-Octafluoropentyl acrylate

Stabilisering: Stabilised with 4-methoxyphenol (MEHQ)

O0318-25G O0318-5G
TCIAO0318-25GEA 1500 NOK
TCIAO0318-25G TCIAO0318-5G
1H,1H,5H-Octafluoropentyl acrylate ≥97.0% (by GC) stabilised
1H,1H,5H-Octafluoropentyl acrylate
Formel: C₈H₆F₈O₂
MW: 286,12 g/mol
Kokepunkt: 84°C
Flash punkt: 72°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00039279
CAS-nummer: 376-84-1
EINECS: 206-816-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f16173aac70c8cb6d8153ccf30ab7381eccabf9f&vid=f16173aac70c8cb6d8153ccf30ab7381eccabf9f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR