5-Chloro-2,4-dinitrobenzoic acid ≥98%
Leverandør: Thermo Scientific
Warning
B25479.06 B25479.14
B25479.06EA 1700 NOK
B25479.06 B25479.14
5-Chloro-2,4-dinitrobenzoic acid ≥98%
5-Chloro-2,4-dinitrobenzoic acid
Purity: 98% CAS No.: 136833-36-8 Molecular Formula: C7H3ClN2O6 Formula Weight: 246.56 Melting Point: 180-182° MDL No.: MFCD00091829
Formel: C₇H₃ClN₂O₆
MW: 246,56 g/mol
Smeltepunkt: 180…182 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00091829
CAS-nummer: 136833-36-8