HPLC columns, NUCLEODUR® HILIC
Leverandør: MACHEREY-NAGEL
760550.30 760553.46 760526.20 760531.40 760533.46 761961.30 760554.30 760526.40 760533.30 760528.20 760525.20 760521.20 760521.30 760521.40 760521.46 760523.20 760523.30 760523.40 760523.46 760526.30 760526.46 760530.20 760530.30 760530.40 760530.46 760531.20 760531.30 760531.46 760532.20 760532.30 760532.40 760532.46 760533.20 760533.40 760534.20 760534.30 760534.40 760534.46 760550.20 760550.40 760550.46 760551.20 760551.30 760551.40 760551.46 760552.20 760552.30 760552.40 760552.46 760553.20 760553.30 760553.40 760554.20 760554.40 760554.46 761960.20 761960.30 761961.20 761962.20 761962.30
MANA760550.30EA 18425 NOK
MANA760550.30 MANA760553.46 MANA760526.20 MANA760531.40 MANA760533.46 MANA761961.30 MANA760554.30 MANA760526.40 MANA760533.30 MANA760528.20 MANA760525.20 MANA760521.20 MANA760521.30 MANA760521.40 MANA760521.46 MANA760523.20 MANA760523.30 MANA760523.40 MANA760523.46 MANA760526.30 MANA760526.46 MANA760530.20 MANA760530.30 MANA760530.40 MANA760530.46 MANA760531.20 MANA760531.30 MANA760531.46 MANA760532.20 MANA760532.30 MANA760532.40 MANA760532.46 MANA760533.20 MANA760533.40 MANA760534.20 MANA760534.30 MANA760534.40 MANA760534.46 MANA760550.20 MANA760550.40 MANA760550.46 MANA760551.20 MANA760551.30 MANA760551.40 MANA760551.46 MANA760552.20 MANA760552.30 MANA760552.40 MANA760552.46 MANA760553.20 MANA760553.30 MANA760553.40 MANA760554.20 MANA760554.40 MANA760554.46 MANA761960.20 MANA761960.30 MANA761961.20 MANA761962.20 MANA761962.30
HPLC columns, NUCLEODUR® HILIC
Kromatografikolonner
NUCLEODUR® HILIC is an ammonium – sulfonic acid modified silica phase for HPLC.
  • High purity spherical silica gel
  • Hydrophilic phase for hydrophilic interaction chromatography (HILIC)
  • Separation mechanism based on partition chromatography between the organic/aqueous mobile and the polar stationary phase
The HILIC phase is recommended for hydrophilic compounds such as organic polar acids and bases, polar natural compounds, nucleosides, oligonucleotides, amino acids, peptides, and water soluble vitamins.
EC guard columns are used for analytical ECMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 7%
Kappet ved endene Nei
pH område 2 - 8,5