1H-Benzimidazole-5-carboxylic acid ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 1H-Benzo[d]imidazole-5-carboxylic acid, Benzimidazole-5-carboxylic acid, 5-Benzimidazolecarboxylic acid

B1763-25G B1763-5G
TCIAB1763-25GEA 2800 NOK
TCIAB1763-25G TCIAB1763-5G
1H-Benzimidazole-5-carboxylic acid ≥97.0% (by titrimetric analysis)
1H-Benzimidazole-5-carboxylic acid
Formel: C₈H₆N₂O₂
MW: 162,15 g/mol
MDL nummer: MFCD00011555
CAS-nummer: 15788-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0284624fb73c847fc2537702b6d1f46350d40670&vid=0284624fb73c847fc2537702b6d1f46350d40670&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR