Okadaic acid
Leverandør: Cayman Chemical
Synonymer: 9,10-deepithio-9,10-didehydroacanthifolicin, OA
10011490-25 10011490-50 10011490-100
CAYM10011490-25EA 1800 NOK
CAYM10011490-25 CAYM10011490-50 CAYM10011490-100
Okadaic acid
Okadaic acid
Formel: C₄₄H₆₈O₁₃
MW: 805.02 g/mol
MDL nummer: MFCD00083455
CAS-nummer: 78111-17-8