1,1-Cyclohexanediacetic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 1,1-Cyclohexanediaceticacid
C1444-25G C1444-5G
TCIAC1444-25GEA 915 NOK
TCIAC1444-25G TCIAC1444-5G
1,1-Cyclohexanediacetic acid ≥98.0% (by GC, titration analysis)
1,1-Cyclohexanediacetic acid
Formel: C₁₀H₁₆O₄
MW: 200,23 g/mol
MDL nummer: MFCD00001519
CAS-nummer: 4355-11-7
EINECS: 224-427-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=836675ecf2b75e5da0407a926f8523b4f3b97eb2&vid=836675ecf2b75e5da0407a926f8523b4f3b97eb2&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 178.0 to 184.0 deg-C
Solubility in Methanol almost transparency