Victoria pure blue bo

Leverandør: TCI

Synonymer: Basic Blue 7, N-(4-((4-(Diethylamino)phenyl)(4-(ethylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium chloride

B1301-25G B1301-250G
TCIAB1301-25GEA 570 NOK
TCIAB1301-25G TCIAB1301-250G
Victoria pure blue bo
Victoria Pure Blue Bo
Formel: C₃₃H₄₀ClN₃
MW: 514,1438 g/mol
MDL nummer: MFCD00011877
CAS-nummer: 2390-60-5
EINECS: 219-232-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4badfcc081722b2a1ca9fa33484d4a05fbb8d39&vid=c4badfcc081722b2a1ca9fa33484d4a05fbb8d39&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to Amber to Dark red powder to crystal
Lambda max. 613.0 to 619.0 nm (H2O)
Absorbance (E1%1cm) min. 1,100 (H2O, 613.0 to 619.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR