2,2'-Biquinoline ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

B0470-1G B0470-5G
TCIAB0470-1GEA 405 NOK
TCIAB0470-1G TCIAB0470-5G
2,2'-Biquinoline ≥98.0% (by HPLC, titration analysis)
2,2'-Biquinoline
Formel: C₁₈H₁₂N₂
MW: 256.31 g/mol
MDL nummer: MFCD00006740
CAS-nummer: 119-91-5
EINECS: 204-357-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c45088a4473fc65d616129dc19de2283e65045de&vid=c45088a4473fc65d616129dc19de2283e65045de&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 192.0 to 197.0 deg-C
Solubility in hot Acetic acid almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR