Bathocuproin ≥95.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

D0711-1G D0711-5G
TCIAD0711-1GEA 1150 NOK
TCIAD0711-1G TCIAD0711-5G
Bathocuproin ≥95.0% (by HPLC, titration analysis)
Bathocuproin
Formel: C₂₆H₂₀N₂
Smeltepunkt: 283°C
MDL nummer: MFCD00004972
CAS-nummer: 4733-39-5
EINECS: 225-240-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99e2fb8dab7c0223f232396c83d5cd7932b54966&vid=99e2fb8dab7c0223f232396c83d5cd7932b54966&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Green powder to crystaline
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR