Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
B22429.06 B22429.14 B22429.22
B22429.06EA 355 NOK
B22429.06 B22429.14 B22429.22
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate ≥98%
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Purity: 98% CAS No.: 5909-24-0 Molecular Formula: C8H9ClN2O2S Formula Weight: 232.69 Storage Temperature: -30°C to -10°C Melting Point: 60-63° MDL No.: MFCD00006085
Formel: C₈H₉ClN₂O₂S
MW: 232,69 g/mol
Smeltepunkt: 60…63 °C
MDL nummer: MFCD00006085
CAS-nummer: 5909-24-0
EINECS: 227-619-0