(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: [(5-Sulphanyl-1,3,4-thiadiazol-2-yl)sulphanyl]acetic acid

M2100-25G M2100-5G
TCIAM2100-25GEA 3075 NOK
TCIAM2100-25G TCIAM2100-5G
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid ≥97.0% (by HPLC, titration analysis)
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid
Formel: C₄H₄N₂O₂S₃
MW: 208,28 g/mol
MDL nummer: MFCD00129970
CAS-nummer: 53723-88-9
EINECS: 258-728-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42aded0cc5744c0a2d628babf64c7feb3503d388&vid=42aded0cc5744c0a2d628babf64c7feb3503d388&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 165.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR