2-Sulphobenzoic acid hydrate ≥98%
Leverandør: Thermo Scientific
Danger
Synonymer: 2-sulphobenzoic acid hydrate
B23381.14 B23381.22
B23381.14EA 570 NOK
B23381.14 B23381.22
2-Sulphobenzoic acid hydrate ≥98%
2-Sulphobenzoic acid hydrate
Purity: 98% CAS No.: 123333-68-6 Molecular Formula: C7H6O5S.xH2O Formula Weight: 202.19(anhy) Melting Point: ca 68-72° Beilstein Reference: 2106852 UN No.: 3261 MDL No.: MFCD00149534
Formel: C₇H₆O₅S·H₂O
MW: 202,19 g/mol (anhy)
Smeltepunkt: ∼ 68…72 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149534
CAS-nummer: 123333-68-6
EINECS: 211-176-5
ADR: 8,II