(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-(+)-Phenyllactic acid
P1981-1G
P1981-5G
TCIAP1981-1GEA
220
NOK
TCIAP1981-1G
TCIAP1981-5G
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)
(R)-(+)-2-Hydroxy-3-phenylpropionic acid
Formel:
C₉H₁₀O₃ MW: 166,1739 g/mol |
MDL nummer:
MFCD00078062 CAS-nummer: 7326-19-4 EINECS: 230-803-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Specific rotation [a]20/D | +20.0 to +22.0 deg (C=2, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...