3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid
B20412.14 B20412.06 B20412.22
B20412.14EA 4950 NOK
B20412.14 B20412.06 B20412.22
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid ≥98%
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid
Purity: 98% CAS No.: 20170-32-5 Molecular Formula: C17H26O3 Formula Weight: 278.40 Melting Point: 172-175° MDL No.: MFCD00017519
Formel: C₁₇H₂₆O₃
MW: 278,40 g/mol
Smeltepunkt: 172…175 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00017519
CAS-nummer: 20170-32-5
EINECS: 243-556-1