N-(3-Aminopropyl)-N-methylaniline ≥96.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: N1-Methyl-N1-phenylpropane-1,3-diamine
A1295-25ML A1295-500ML
TCIAA1295-25MLEA 865 NOK
TCIAA1295-25ML TCIAA1295-500ML
N-(3-Aminopropyl)-N-methylaniline ≥96.0% (by GC, titration analysis)
N-(3-Aminopropyl)-N-methylaniline
Formel: C₁₀H₁₆N₂
MW: 164,250 g/mol
Kokepunkt: 172°C
MDL nummer: MFCD00191602
CAS-nummer: 53485-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08487b0bf560d546f1bceba606fb0d56d3f8b2d6&vid=08487b0bf560d546f1bceba606fb0d56d3f8b2d6&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %