2-Nitroterephthalic acid 1-methyl ester ≥97%
Leverandør: Thermo Scientific
Warning
Synonymer: 1-Methyl 2-nitroterephthalate
H27121.06 H27121.14
H27121.06EA 330 NOK
H27121.06 H27121.14
2-Nitroterephthalic acid 1-methyl ester ≥97%
2-Nitroterephthalic acid 1-methyl ester
Purity: 97% CAS No.: 35092-89-8 Molecular Formula: C9H7NO6 Formula Weight: 225.17 Melting Point: 173-178° MDL No.: MFCD00024510
Formel: C₉H₇NO₆
MW: 225,17 g/mol
Smeltepunkt: 173…178 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00024510
CAS-nummer: 35092-89-8
EINECS: 252-360-5