6-Methylpicolinic acid 95%
Leverandør: Thermo Scientific
Warning
Synonymer: 6-Methyl-2-pyridinecarboxylic acid, 6-Methylpyridine-2-carboxylic acid
H26302.03 H26302.06
H26302.03EA 1075 NOK
H26302.03 H26302.06
6-Methylpicolinic acid 95%
6-Methylpicolinic acid
Purity: 95% CAS No.: 934-60-1 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Odour: Odourless Melting Point: 127-131° Boiling Point: 100°/4.5mm MDL No.: MFCD00023481
Formel: C₇H₇NO₂
MW: 137,14 g/mol
Kokepunkt: 100 °C (4,5 mmHg)
Smeltepunkt: 127…131 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00023481
CAS-nummer: 934-60-1
EINECS: 213-287-4