N-(2-Cyanoethyl)-N-methylaniline ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 3-(Methyl(phenyl)amino)propanenitrile

C0872-25G C0872-500G
TCIAC0872-25GEA 245 NOK
TCIAC0872-25G TCIAC0872-500G
N-(2-Cyanoethyl)-N-methylaniline ≥98.0% (by titrimetric analysis)
N-(2-Cyanoethyl)-N-methylaniline
Formel: C₁₀H₁₂N₂
MW: 160,2200 g/mol
Kokepunkt: 186°C
MDL nummer: MFCD00019856
CAS-nummer: 94-34-8
EINECS: 202-325-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b59e27cc02e48ea95e53c6aa9366c9ce5cfea9c5&vid=b59e27cc02e48ea95e53c6aa9366c9ce5cfea9c5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR