8-Amino-2-methylquinoline ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 2-Methylquinolin-8-amine, 8-Aminoquinaldine

A0416-1G A0416-5G
TCIAA0416-1GEA 785 NOK
TCIAA0416-1G TCIAA0416-5G
8-Amino-2-methylquinoline ≥98.0% (by titrimetric analysis)
8-Amino-2-methylquinoline
Formel: C₁₀H₁₀N₂
MW: 158,1998 g/mol
MDL nummer: MFCD00023998
CAS-nummer: 18978-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd599debea9a3afbbdbc5c33b17a4daaeec271d8&vid=fd599debea9a3afbbdbc5c33b17a4daaeec271d8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Amber to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR