Cyclobutyl methyl ketone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Acetylcyclobutane, Ketone cyclobutyl methyl, 1-Cyclobutylethan-1-one, 1-Cyclobutylethanone, Cyclobutyl Methyl Ketone

C1034-1ML C1034-5ML
TCIAC1034-1MLEA 1375 NOK
TCIAC1034-1ML TCIAC1034-5ML
Cyclobutyl methyl ketone ≥98.0% (by GC)
Cyclobutyl methyl ketone
Formel: C₆H₁₀O
MW: 98,15 g/mol
Kokepunkt: 137°C
Flash punkt: 28°C
MDL nummer: MFCD00037148
CAS-nummer: 3019-25-8
EINECS: 221-163-6
FN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c3ab3d4eded953808d6bc91488e6f5ac1470798&vid=9c3ab3d4eded953808d6bc91488e6f5ac1470798&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR