1-Aminopiperidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

A2241-25G A2241-5G
TCIAA2241-25GEA 1875 NOK
TCIAA2241-25G TCIAA2241-5G
1-Aminopiperidine ≥98.0% (by GC, titration analysis)
1-Aminopiperidine
Formel: C₅H₁₂N₂
MW: 100,16 g/mol
Kokepunkt: 146°C
Flash punkt: 36°C
MDL nummer: MFCD00006489
CAS-nummer: 2213-43-6
EINECS: 218-666-8
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27865d3e79444ee12815597215e583073b652d1c&vid=27865d3e79444ee12815597215e583073b652d1c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR