5-Bromo-2-hydroxy-3-picoline ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Bromo-3-methylpyridin-2-ol, 5-Bromo-2-hydroxy-3-methylpyridine, 5-Bromo-3-methylpyridin-2(1H)-one

B2797-1G B2797-5G
TCIAB2797-1GEA 300 NOK
TCIAB2797-1G TCIAB2797-5G
5-Bromo-2-hydroxy-3-picoline ≥98.0% (by GC)
5-Bromo-2-hydroxy-3-picoline
Formel: C₆H₆BrNO
MW: 188,02 g/mol
MDL nummer: MFCD03427657
CAS-nummer: 89488-30-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b88bb3685523bb1826d1da34119b81c51308cbc&vid=7b88bb3685523bb1826d1da34119b81c51308cbc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 170.0 to 174.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR