(R)-(-)-Ethyl-3-piperidinecarboxylate ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Ethyl (R)-(-)-3-Piperidinecarboxylate, (R)-Ethyl piperidine-3-carboxylate
N0655-25G
N0655-5G
TCIAN0655-25GEA
5550
NOK
TCIAN0655-25G
TCIAN0655-5G
(R)-(-)-Ethyl-3-piperidinecarboxylate ≥98.0% (by GC)
(R)-(-)-Ethyl-3-piperidinecarboxylate
Formel:
C₈H₁₅NO₂ MW: 157,2102 g/mol Kokepunkt: 110°C Flash punkt: 90°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD03093637 CAS-nummer: 25137-01-3 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -1.0 to -2.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...