Ethyl-2-piperidinecarboxylate ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Ethyl 2-Piperidinecarboxylate, Ethyl pipecolinate
E0837-25G
E0837-5G
TCIAE0837-25GEA
2075
NOK
TCIAE0837-25G
TCIAE0837-5G
Ethyl-2-piperidinecarboxylate ≥98.0% (by GC, titration analysis)
Ethyl-2-piperidinecarboxylate
Formel:
C₈H₁₅NO₂ MW: 157,21 g/mol Kokepunkt: 217°C Flash punkt: 46°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00005980 CAS-nummer: 15862-72-3 EINECS: 239-990-6 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...