3,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
Synonymer:
Dihydroxy(3,4-dimethylphenyl)borane, (3,4-Dimethylphenyl)metaboric acid, 3,4-Dimethylbenzeneboronic acid
D3110-5G
D3110-1G
TCIAD3110-5GEA
2025
NOK
TCIAD3110-5G
TCIAD3110-1G
3,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
3,4-Dimethylphenylboronic acid
Formel:
C₈H₁₁BO₂ MW: 149,98 g/mol |
MDL nummer:
MFCD01009694 CAS-nummer: 55499-43-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...